Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MNQSRSRSD GGSEETLPQD HNHHENERRW QQERLHREEA YYQFINELND EDYRLMRDHN LLGTPGEITS EELQQRLDGV KEQLASQPDL RDGTNYRDSE VPRESSHEDS LLEWLNTFRR TGNATRSGQN GNQTWRAVSR TNPNNGEFRF SLEIHVNHEN RGFEIHGEDY TDIPLSDSNR DHTANRQQRS TSPVARRTRS QTSVNFNGSS SNIPRTRLAS RGQNPAEGSF STLGRLRNGI GGAAGIPRAN ASRTNFSSHT NQSGGSELRQ REGQRFGAAH VWENGARSNV TVRNTNQRLE P (1-300 aa encoded by BC034688)
The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686P0776; E3 ubiquitin-protein ligase RNF6; ring finger protein (C3H2C3 type) 6; RING-H2 protein RNF-6; RLIM-like protein
Gene Aliases: 1200013I08Rik; 5730419H05Rik; AA537053; RNF6; SPG2
UniProt ID: (Human) Q9Y252, (Mouse) Q9DBU5
Entrez Gene ID: (Human) 6049, (Mouse) 74132
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support