Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX24541-PEP)..
This antibody is specific for GNIP / TRIM7.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1, a B-box type 2, and a coiled-coil region. The protein localizes to both the nucleus and the cytoplasm, and may represent a participant in the initiation of glycogen synthesis. Multiple transcript variants have been found for this gene, and some of them encode the same isoform.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM7; Glycogenin-interacting protein; RING finger protein 90; tripartite motif protein 7; tripartite motif protein TRIM7; Tripartite motif-containing protein 7
Gene Aliases: AI790312; GNIP; RNF90; TRIM7
UniProt ID: (Human) Q9C029, (Mouse) Q923T7
Entrez Gene ID: (Human) 81786, (Mouse) 94089
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support