Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LEVFYQTQGR LHPNLRRAIQ QILSQGLGSS TEPASEPSTE LGKAEADTDS DAQALLLGDE APSSAISLRD VNVSA
Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
RNPEPL1 has broad specificity aminopeptidase which preferentially hydrolyzes an N-terminal methionine, citrulline or glutamine.".
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aminopeptidase B-like 1; Aminopeptidase RNPEPL1; argininyl aminopeptidase-like 1; arginyl aminopeptidase (aminopeptidase B)-like 1; Arginyl aminopeptidase-like 1; Methionyl aminopeptidase; RNPEP-like 1
Gene Aliases: RNPEPL1
UniProt ID: (Human) Q9HAU8
Entrez Gene ID: (Human) 57140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support