Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: Ribonuclease A, mouse pancreas
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rib 1; ribonuclease 1, pancreatic; ribonuclease A (pancreatic); Ribonuclease BS-1; Ribonuclease pancreatic; RNase 1; RNase A; RNAseA; S-RNase; Seminal ribonuclease; seminal RNase
Gene Aliases: AI574248; BOS_10503; BOS_10505; Rib-1; Rib1; RNASE1; RNS1; SRN
UniProt ID: (Bovine) P00669, (Bovine) P61823
Entrez Gene ID: (Bovine) 280930, (Bovine) 282340, (Mouse) 19752
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support