Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GSDELLSSGI INGPFTMNSS TPSTANGNDS KKFKRDRPPC SPSRVLHLRK IPCDVTEAEI ISLGLPFGKV TNLLMLKGKS QAFLEMASEE AAVTMVNYY
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fission yeast differentiation regulator; Polypyrimidine tract-binding protein 3; regulator of differentiation (in S. pombe) 1; Regulator of differentiation 1; Rod1; ROD1 regulator of differentiation 1
Gene Aliases: 5830471K22Rik; AA407443; AI462022; AW107884; C86549; PTBP3; ROD1
UniProt ID: (Human) O95758, (Mouse) Q8BHD7, (Rat) Q9Z118
Entrez Gene ID: (Human) 9991, (Mouse) 230257, (Rat) 83515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support