Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
ROR gamma is a 560 amino acid novel DNA-binding transcription factor belonging to the ROR/RZR orphan nuclear receptor subfamily. Reports suggest thatROR gammais a ligand activated transcription factor and may be essential for lymphoid organogenesis (lymph nodes and Peyer's patches) and may play an important regulatory role in controlling apoptosis during thymopoiesis. ROR gamma inhibits the expression of Fas ligand and IL2 and directs the differentiation program of proinflammatory IL-17+ T helper cells. ROR gamma shares 50 to 51% sequence identity with RORA and RORB and is 88% identical to mouse ROR gamma. It is highly expressed in skeletal muscle and at lower levels in various other tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma
Gene Aliases: IMD42; NR1F3; RORC; RORG; RZR-GAMMA; RZRG; TOR
UniProt ID: (Human) P51449
Entrez Gene ID: (Human) 6097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support