Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total RPL27A.
Rpl27a gene ontology annotations related to this gene include translation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L27a; FLJ43464; L29; Large ribosomal subunit protein uL15; MGC10850; MGC17878; MGC87238; ribosomal protein L29 homolog
Gene Aliases: L27'; L27A; L29; RPL27A
UniProt ID: (Human) P46776, (Mouse) P14115, (Rat) P18445
Entrez Gene ID: (Human) 6157, (Mouse) 26451, (Rat) 293418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support