Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: GAAPIIDVVR SGYYKVLGKG KLPKQPVIVK AKFFSRRAEE KIKSVGGACV
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Yeast: 85%; Zebrafish: 93%
Rpl27a gene ontology annotations related to this gene include translation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L27a; FLJ43464; Large ribosomal subunit protein uL15; MGC10850; MGC17878; MGC87238
Gene Aliases: L27A; RPL27A
UniProt ID: (Human) P46776
Entrez Gene ID: (Human) 6157
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support