Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGRVIRGQR KGAGSVFRAH VKHRKGAARL RAVDFAERHG YIKGIVKDII HDPGRGAPLA KVVFRDPYRF KKRTELFIAA EGIHTGQFVY CGKKAQLNVG NVLPVGTMPE GTIVCCLEEK PGDRGKLARA SGNYATVISH NPETKKTRVK LPSGSKKVIS SANRAVVGVV AGGGRIDKPI LKAGRAYHKY KAKRNCWPRV RGVAMNPVEH PFGGGNHQHI GKPSTIRRDA PAGRKVGLIA ARRTGRLRGT KTVQEKEN (1-257 aa encoded by BC012197)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L2P family of ribosomal proteins. It is located in the cytoplasm. In rat, the protein associates with the 5. 8S rRNA, very likely participates in the binding of aminoacyl-tRNA, and is a constituent of the elongation factor 2-binding site at the ribosomal subunit interface. Alternatively spliced transcript variants encoding the same protein exist. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L8; Large ribosomal subunit protein uL2
Gene Aliases: L8; RPL8
UniProt ID: (Human) P62917, (Rat) P62919, (Mouse) P62918
Entrez Gene ID: (Human) 6132, (Rat) 26962, (Mouse) 26961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support