Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RPLP1 Antibodies

          Invitrogen

          RPLP1 Monoclonal Antibody (1G10)

          1 Reference
          View all (8) RPLP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RPLP1 Monoclonal Antibody (1G10)

          • Antibody Testing Data (3)
          RPLP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RPLP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RPLP1 Antibody (MA5-24411) in ICC/IF

          Immunofluorescent analysis of RPLP1 in HeLa cell lysates. Samples were probed using a Mouse Monoclonal RPLP1 (1G10) Antibody (Product # MA5-24411) at 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RPLP1 Antibody in Immunocytochemistry (ICC/IF)
          RPLP1 Antibody in Western Blot (WB)
          RPLP1 Antibody in ELISA (ELISA)

          Product Details

          MA5-24411

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -

          ELISA (ELISA)

          1 ng/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1G10

          Immunogen

          RPLP1 (NP_000994.1, 1 a.a. ~ 114 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.49 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2633383

          Product Specific Information

          Peptide Sequence: MASVSELACI YSALILHDDE VTVTEDKINA LIKAAGVNVE PFWPGLFAKA LANVNIGSLI CNVGAGGPAP AAGAAPAGGP APSTAAAPAE EKKVEAKKEE SEESDDDMGF GLFD

          Target Information

          Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; L RPP1; Large ribosomal subunit protein P1; ribosomal protein, large, P1

          View more View less

          Gene Aliases: LP1; P1; RPLP1; RPP1; RRP1

          View more View less

          UniProt ID: (Human) P05386

          View more View less

          Entrez Gene ID: (Human) 6176

          View more View less

          Function(s)
          structural constituent of ribosome protein binding protein kinase activator activity large ribosomal subunit rRNA binding
          Process(es)
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay cytoplasmic translation rRNA processing translation translational initiation translational elongation SRP-dependent cotranslational protein targeting to membrane viral transcription activation of protein kinase activity positive regulation of protein kinase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline