Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PAATYERVVY KNPSEYHYMK VCLEFQDCGV GLNAAQFKQL LISAVKDLFG EVDAALPLDI LTYEEKTLSA ILRICSSGLV K
Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
This gene encodes a subunit of ribonuclease P and has 3' to 5' exoribonuclease activity. Transcripts for this gene are bicistronic and include a conserved downstream open reading frame for the hydroxyacyl-thioester dehydratase type 2 (HTD2) gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; Hydroxyacyl-thioester dehydratase type 2, mitochondrial; Ribonuclease P protein subunit p14; ribonuclease P/MRP 14kDa subunit
Gene Aliases: HsHTD2; P14; RPP14
UniProt ID: (Human) O95059
Entrez Gene ID: (Human) 11102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support