Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15 and may have a role in cell cycle regulation (Liu et al., 2002 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDKN2B-related protein; cyclin-dependent kinase 2B-inhibitor-related protein; Cyclin-dependent kinase inhibitor 2B-related protein; Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein); p15INK4B-related protein; Regulation of nuclear pre-mRNA domain-containing protein 1A
Gene Aliases: HsT3101; P15RS; RPRD1A
UniProt ID: (Human) Q96P16
Entrez Gene ID: (Human) 55197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support