Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VTKMKLQRTI VIRRDYLHYI RKYNRFEKRH KNMSVHLSPC FRDVQIGDIV TVGECRPLSK TVRFNVLEV
Antigen sequence identity: Mouse 97%; Rat 97%
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S17P family of ribosomal proteins. It is located in the cytoplasm. The gene product of the E. coli ortholog (ribosomal protein S17) is thought to be involved in the recognition of termination codons. This gene is co-transcribed with a small nucleolar RNA gene, which is located in its third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S11; RPS 11; Small ribosomal subunit protein uS17
Gene Aliases: RPS11; S11
UniProt ID: (Human) P62280
Entrez Gene ID: (Human) 6205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support