Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPGSVENLS IVYRSRDFLV VNKHWDVRID SKAWRETLTL QKQLRYRFPE LADPDTCYGF RFCHQLDFST SGALCVALNK AAAGSAYRCF KERRVTKAYL ALLRGHIQES RVTISHAIGR NSTEGRAHTM CIEGSQGCEN PKPSLTDLVV LEHGLYAGDP VSKVLLKPLT GRTHQLRVHC SALGHPVVGD LTYGEVSGRE DRPFRMMLHA FYLRIPTDTE CVEVCTPDPF LPSLDACWSP HTLLQSLDQL VQALRATPDP DPEDRGPRPG SPSALLPGPG RPPPPPTKPP ETEAQRGPCL QWLSEWTLEP DS
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribosomal large subunit pseudouridine synthase C-like protein; RNA pseudouridylate synthase domain-containing protein 1
Gene Aliases: C16orf40; RLUCL; RPUSD1
UniProt ID: (Human) Q9UJJ7
Entrez Gene ID: (Human) 113000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support