Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVSRVQLPPE IQLAQRLAGN EQVTRDRAVR KLRKYIVART QRAAGGFTHD ELLKVWKGLF YCMWMQDKPL LQEELGRTIS QLVHAFQTTE AQHLFLQAFW QTMNREWTGI DRLRLDKFYM LMRMVLNESL KVLKMQGWEE RQIEELLELL MTEILHPSSQ APNGVKSHFI EIFLEELTKV GAEELTADQN LKFIDPFCRI AARTKDSLVL NNITRGIFET IVEQAPLAIE DLLNELDTQD EEVASDSDES SEGGERGDAL SQKRSEKPPA GSICRAEPEA GEEQAGDDRD SGGPVLQFDY EAVANRLFEM ASRQSTPSQN RKRLYKVIRK LQDLAGGIFP EDEIPEKACR RLLEGRRQKK TKKQKRLLRL QQERGKGEKE PPSPGMERKR SRRRGVGADP EARAEAGEQP GTAERALLRD QPRGRGQRGA RQRRRTPRPL TSARAKAANV QEPEKKKKRR E
The protein encoded by this gene is the putative homolog of the yeast ribosomal RNA processing protein RRP1. The encoded protein is involved in the late stages of nucleologenesis at the end of mitosis, and may be required for the generation of 28S rRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NNP-1; Nnp1 homolog, nucleolar protein; Novel nuclear protein 1; Nucleolar protein Nop52; protein NNP-1; ribosomal RNA processing 1 homolog; Ribosomal RNA processing protein 1 homolog A; RRP1-like protein
Gene Aliases: D21S2056E; NNP-1; NNP1; NOP52; RRP1; RRP1A
UniProt ID: (Human) P56182
Entrez Gene ID: (Human) 8568
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support