Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MKMVTGAVAS VLEDEATDTS DSEGSCGSEK DHFYSDDDAI EADSEGDAEP CDKENENDGE SSVGTNMGWA DAMAKVLNKK TPESKPTILV KNKKLEKEKE KLKQERLEKI KQRDKRLEWE MMCRVKPDVV QDKETERNLQ RIATRGVVQL FNAVQKHQKN VDEKVKEAGS SMRKRAKLIS TVSKKDFISV LRGMDGSTNE TASSRKKPKA KQTEVKSEEG PGWTILRDDF MMGASMKDWD KESDGPDDSR PESASDSDT
This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribosomal RNA-processing protein 15; RRP15-like protein
Gene Aliases: CGI-115; KIAA0507; RRP15
UniProt ID: (Human) Q9Y3B9
Entrez Gene ID: (Human) 51018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support