Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: rat testis tissue, rat PC-12 whole cell.
ATP-dependent DNA helicase implicated in telomere-length regulation, DNA repair and the maintenance of genomic stability. Acts as an anti-recombinase to counteract toxic recombination and limit crossover during meiosis. Regulates meiotic recombination and crossover homeostasis by physically dissociating strand invasion events and thereby promotes noncrossover repair by meiotic synthesis dependent strand annealing (SDSA) as well as disassembly of D loop recombination intermediates. Also disassembles T loops and prevents telomere fragility by counteracting telomeric G4-DNA structures, which together ensure the dynamics and stability of the telomere.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAH helicase; helicase-like protein NHL; Novel helicase-like; Regulator of telomere elongation helicase 1; regulator of telomere elongation helicase 1 {ECO:0000255|HAMAP-Rule:MF_03065}; regulator of telomere length; RTEL1
Gene Aliases: AI451565; AW540478; C20orf41; DKCA4; DKCB5; KIAA1088; NHL; PFBMFT3; RGD1306721; RTEL; RTEL1
UniProt ID: (Human) B4DYM3, (Rat) Q5RJZ1, (Mouse) Q0VGM9
Entrez Gene ID: (Human) 51750, (Rat) 362288, (Mouse) 269400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support