Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen sequence: MAEPSAATQS HSISSSSFGA EPSAPGGGGS PGACPALGTK SCSSSCAVHD LIFWRDVKKT GFVFGTTLIM LLSLAAFSVI SVVSYLILAL LSVTISFRIY KSVIQAVQKS EEGHPFKAYL DVDITLSSEA FHNYMNAAMV HINRALKLII RLFLVEDLVD SLKLAVFMWL MTYVGAVFNG ITLLILAELL IFSVPIVYEK YKTQIDHYVG IARDQTKSIV EKIQAKLPGI AKKKAE
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASY interacting protein; HAP; Homolog of ASY protein; Neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSP-like protein 2; NSP-like protein II; Reticulon-3
Gene Aliases: ASYIP; HAP; NSPL2; NSPLII; RTN3; RTN3-A1
UniProt ID: (Human) O95197
Entrez Gene ID: (Human) 10313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support