Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
RTP3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3CxxC-type zinc finger protein 3; receptor (chemosensory) transporter protein 3; receptor transporting protein 3; Receptor-transporting protein 3; Transmembrane protein 7; zinc finger, 3CxxC-type 3
Gene Aliases: LTM1; RTP3; TMEM7; Z3CXXC3
UniProt ID: (Human) Q9BQQ7, (Mouse) Q5QGU6
Entrez Gene ID: (Human) 83597, (Mouse) 235636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support