Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ITSFEGKTNQ VMSSMKQMEE RLQHSERARQ GAEERSHKLQ QELGGRIGAL QLQLSQLHEQ CSSLEKELKS EKEQRQALQR ELQHEKDTSS LLRMELQQV
This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FYVE-finger protein EIP1; La-binding protein 1; Rab4-interacting protein; RUN and FYVE domain-containing protein 1; Zinc finger FYVE domain-containing protein 12
Gene Aliases: RABIP4; RUFY1; ZFYVE12
UniProt ID: (Human) Q96T51
Entrez Gene ID: (Human) 80230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support