Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: NCI-H460, SKOV3, HepG2, 293T, Mouse brain, Mouse spleen, Mouse lung, Rat brain, Rat lung. The target is usually found in the following locations: Nucleus.
Immunogen sequence: IDFSMYLKNE EDIGNKERNV QIAAILDQKN YVEELNRQLN STVSSLHSRV DSLEKSNTKL IEELAIAKNN IIKLQEENHQ LRSENKLILM KTQQHLEVTK VDVETELQTY KHSRQGLDEM YNEARRQLRD ESQLRQDVEN ELAVQVSMKH EIELAMKLLE KDIHEKQDTL IGLRQQLEEV KAINIEMYQK LQGSEDGLKE KNEIIARLEE KTNKITAAMR QLEQSDNDLL TQTRTIAMSL VKCASSDTQD QYKLVKDISF
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen MU-RMS-40.17; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein
Gene Aliases: 2610111M19Rik; AI844453; Denn; KIAA1537; LZ-FYVE; RABIP4R; RUFY2; ZFYVE13
UniProt ID: (Human) Q8WXA3, (Mouse) Q8R4C2
Entrez Gene ID: (Human) 55680, (Rat) 690777, (Mouse) 70432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support