Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MDEERSSMLP TMAAGPNSIL FAINIDNKDL NGQSKFAPTV SDLLKESTQN VTLLKESTQG VSSVFREITA SSAISILIKP EQETDPLPVV SRNVSADAKC KKERKKKKQV TNIISFDDEE DEQNSGDMFK KTPGAGESSE DNSDHSSVNI MSAFESPFGP NSNGSQSSNS WKIDSLSLNR EFGYQKLDVK SIDDEDVDEN EDDVYGNSSG RKHRGHSESP EKNGAHSVTQ AGVQWHDLGS LQPLPPGFR
SNX29 also known as RUNDC2A, is a 375 amino protein that contains one RUN domain. RUNDC2A is encoded by a gene that maps to human chromosome 16p13.13, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RUN domain containing 2A; RUN domain-containing protein 2A; SODD; Sorting nexin-29
Gene Aliases: A-388D4.1; RUNDC2A; SNX29
UniProt ID: (Human) Q8TEQ0
Entrez Gene ID: (Human) 92017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support