Invitrogen
The PA1-73078 antibody reacts with Rubella virus virions.
PA1-73078 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications.
The PA1-73078 antibody was raised against Rubella virus strain HPV77.
Rubella virus was first isolated in 1962 by Parkman and Weller. Rubella virus is classified as a togavirus, genus Rubivirus. It is most closely related to group A arboviruses, such as Eastern and Western Equine Encephalitis viruses. It is an enveloped RNA virus, with a single antigenic type that does not cross-react with other members of the togavirus group. Rubella virus is relatively unstable and is inactivated by lipid solvents, trypsin, formalin, ultraviolet light, low pH, heat, and amantadine.