Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: ERRLGKVIPV LQTKTRTNVP TFSQSNLEQQ KQLYLRSVIA HIEDPEDTNQ
Sequence homology: Cow: 85%; Dog: 85%; Horse: 86%; Human: 100%; Mouse: 85%; Pig: 85%; Rabbit: 77%; Rat: 85%
The specific function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: S100P binding protein 1; S100P-binding protein; S100P-binding protein Riken
Gene Aliases: S100PBP; S100PBPR
UniProt ID: (Human) Q96BU1
Entrez Gene ID: (Human) 64766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support