Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: METWSVEQV CSWLVEKNLG ELVHRFQEEE VSGAALLALN DRMVQQLVKK IGHQAVLMDL IKKYKQNTQG LKSPENPKKA ALVMQTEAAR DYRDKESSSP ARHGEQMPSF YPAENLDNGL IDQRVLKQRR NVKQILARSK ALQWTKSYVL PEFPYDVKCM LAEQKCPDHS MRIRIIEFLQ ADMTKYLEGS LYPSTQQYND VVNALLQAHP FLDEDGCGFA GV (1-221 aa encoded by BC029851)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SAM domain-containing protein 3; Sterile alpha motif domain-containing protein 3
Gene Aliases: Gm623; SAMD3
UniProt ID: (Human) Q8N6K7, (Mouse) Q8C4H2
Entrez Gene ID: (Human) 154075, (Mouse) 268288, (Rat) 685940
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support