Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NVRDHDASV YLRLQGDALS VGGYEANPIF WEEVSDKFAF GLFDLDWEVF TQHIEGAINR VPVLEKTGIK STVCGPESFT PDHKPLMGEA PELRGFFLGC GFNSAGMMLG GGCGQELAHW IIH (309-430 aa encoded by BC136364)
The function remains known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPR-2; dimethylglycine dehydrogenase-like 1; FLJ36475; Sarcosine dehydrogenase, mitochondrial; SarDH; sardh {ECO:0000312|RGD:621125}
Gene Aliases: BPR-2; DMGDHL1; SAR; SARD; SARDH; SDH
UniProt ID: (Human) Q9UL12, (Mouse) Q99LB7, (Rat) Q64380
Entrez Gene ID: (Human) 1757, (Mouse) 192166, (Rat) 114123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support