Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Aliquoting is unnecessary for -20°C storage.
The coronavirus nonstructural protein 13 (nsp13) is one of the SARS-Coronavirus replicase cleaving products encoded by ORF1b. Nsp13 is thought to be part of the viral replication complex, which is associated with intracellular membranes. Nsp13 contains a C-terminal NTPase/Helicase domain and an N-terminal putative zinc-binding motif.
Protein Aliases: COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope