FIGURE: 1 / 3
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
Coronaviruses are positive-sense RNA viruses that generate double-stranded RNA (dsRNA) intermediates during replication, yet evade detection by host innate immune sensors. SARS-CoV-2 NSP5 is coronavirus nonstructural protein 15 (nsp15), an endoribonuclease has been reported to be involved in evasion of dsRNA sensors.
Protein Aliases: COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope