Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MKLVTIFLLV TISLCSYSAT AFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLV
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pneumo secretory protein 1; PnSP-1; Secretoglobin family 3A member 2; secretoglobin, family 3A, member 2; UGRP1pnSP-1; Uteroglobin-related protein 1
Gene Aliases: LU103; pnSP-1; PNSP1; SCGB3A2; UGRP1; UNQ566/PRO1128
UniProt ID: (Human) Q96PL1
Entrez Gene ID: (Human) 117156
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support