Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This antibody recognizes intracellular part of human transmembrane adaptor SCIMP. This protein of 17 kDa predicted Mw migrates as a 22 kDa band on SDS PAGE.
SCIMP (SLP adaptor and Csk interacting membrane protein), also known as Nvl, is a palmitoylated transmembrane adaptor protein expressed in professional antigen presenting cells, most prominently in the lymph nodes and spleen. It is associated with tetraspanin-enriched microdomains (together with MHC II). There is a close relationship between SCIMP and tyrosinkinase Lyn, which is constitutively bound to it by its SH3 domain. After MHC II-mediated stimulation in the immunological synapse SCIMP becomes phosphorylated at several tyrosine residues and provides docking sites for Grb2 and SLP65 or SLP76 adaptors transducing the signal downstream, as well as for the kinase Csk with modulatory roles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DTFT5783; SLP adapter and CSK-interacting membrane protein; SLP65/SLP76, Csk-interacting membrane protein; transmembrane protein C17orf87
Gene Aliases: C17orf87; SCIMP; UNQ5783; UNQ5783/PRO16090
UniProt ID: (Human) Q6UWF3
Entrez Gene ID: (Human) 388325
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support