Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSSLYPSLED LKVDQAIQAQ VRASPKMPAL PVQATAISPP PVLYPNLAEL ENYMGLSLSS QEVQESLLQI PEGDSTAVSG PGPGQMVAPV TGYSLGVRRA EIKPGVREIH LCKDERGKTG LRLRKVDQGL FVQLVQANTP ASLVGLRFGD QLLQIDGRDC AGWSSHKAHQ VVKKASGDKI VVVVRDRPFQ RTVTMHKDSM GHVGFVIKKG KIVSLVKGSS AARNGLLTNH YVCEVDGQNV IGLKDKKIME ILATAGNVVT LTIIPSVIYE HMVKKLPPVL LHHTMDHSIP DA
SDCBP2 contains 2 PDZ (DHR) domains. The function of SDCBP2 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Similar to TACIP18; SITAC; syndecan binding protein (syntenin) 2; Syndecan-binding protein 2; Syntenin-2
Gene Aliases: SDCBP2; SITAC; SITAC18; ST-2
UniProt ID: (Human) Q9H190
Entrez Gene ID: (Human) 27111
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support