Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SEC13 Antibodies

          Proteintech

          SEC13 Polyclonal Antibody

          13 References
          View all (12) SEC13 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SEC13 Polyclonal Antibody

          • Antibody Testing Data (23)
          SEC13 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SEC13 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SEC13 Antibody (15397-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 15397-1-AP (SEC13 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SEC13 Antibody in Immunocytochemistry (ICC/IF)
          SEC13 Antibody in Immunocytochemistry (ICC/IF)
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC13 Antibody in Western Blot (WB)
          SEC13 Antibody in Western Blot (WB)
          SEC13 Antibody in Immunoprecipitation (IP)

          Product Details

          15397-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SEC13 Fusion Protein Ag7610 (1-325 aa encoded by BC002634)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGKMVSVIN TVDTSHEDMI HDAQMDYYGT RLATCSSDRS VKIFDVRNGG QILIADLRGH EGPVWQVAWA HPMYGNILAS CSYDRKVIIW REENGTWEKS HEHAGHDSSV NSVCWAPHDY GLILACGSSD GAISLLTYTG EGQWEVKKIN NAHTIGCNAV SWAPAVVPGS LIDHPSGQKP NYIKRFASGG CDNLIKLWKE EEDGQWKEEQ KLEAHSDWVR DVAWAPSIGL PTSTIASCSQ DGRVFIWTCD DASSNTWSPK LLHKFNDVVW HVSWSITANI LAVSGGDNKV TLWKESVDGQ WVCISDVNKG QGSVSASVTE GQQNEQ (1-325 aa encoded by B C002634)

          Target Information

          The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GATOR2 complex protein SEC13; Protein SEC13 homolog; SEC13 homolog; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13 related; SEC13-like 1; SEC13-like 1 isoform; SEC13-like protein 1; SEC13-related protein

          View more View less

          Gene Aliases: 1110003H02Rik; D3S1231E; npp-20; SEC13; SEC13A; SEC13L1; SEC13R

          View more View less

          UniProt ID: (Human) P55735, (Mouse) Q9D1M0, (Rat) Q5XFW8

          View more View less

          Entrez Gene ID: (Human) 6396, (Mouse) 110379, (Rat) 297522

          View more View less

          Function(s)
          protein binding
          Process(es)
          antigen processing and presentation of peptide antigen via MHC class I intracellular protein transport sister chromatid cohesion antigen processing and presentation of exogenous peptide antigen via MHC class II positive regulation of TOR signaling COPII vesicle coating mRNA transport transport ER to Golgi vesicle-mediated transport protein transport vesicle-mediated transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0