Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein transport protein Sec24D; SEC24 family, member D; SEC24 related gene family, member D; SEC24-related protein D
Gene Aliases: 2310020L09Rik; CLCRP2; Gm1349; KIAA0755; SEC24D
UniProt ID: (Human) O94855
Entrez Gene ID: (Human) 9871, (Mouse) 69608, (Rat) 310843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support