Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: HARCQPGGGP PSPPPGIPGQ PLPSPTRLHL GGGRNSNANG YVRLQLGGED RGGLGHPLPE LADELRRKLQ QRQPLPDSNP EESSV; Positive Samples: MCF7, BT-474, Mouse heart; Cellular Location: Cell junction, Cytoplasmic vesicle, Single-pass type I membrane protein, postsynaptic cell membrane, postsynaptic density, secretory vesicle, synapse, synaptic vesicle membrane
SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20369; M-Sema F; MGC126382; MGC126383; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; Semaphorin-4C; Semaphorin-C-like 1
Gene Aliases: AI426163; KIAA1739; M-Sema F; M-SEMA-F; S4c; sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; UNQ5855/PRO34487
UniProt ID: (Human) Q9C0C4, (Mouse) Q64151
Entrez Gene ID: (Human) 54910, (Mouse) 20353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support