Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MRVFLLCAYI LLLMVSQLRA VSFPEDDEPL NTVDYHYSRQ YPVFRGRPSG NESQHRLDFQ LMLKIRDTLY IAGRDQVYTV NLNEMPKTEV IPNKKLTWRS RQQDRENCAM KGKHKDECHN FIKVFVPRND EMVFVCGTNA FNPMCRYYRL STLEYDGEEI SGLARCPFDA RQTNVALFAD GKLYSATVAD FLASDAVIYR SMGDGSALRT IKYDSKWIKE PHFLHAIEYG NYVYFFFREI AVEHNNLGKA VYSRVARICK NDMGGSQRVL EKHWTSFLKA RLNCSVPGDS FFYFDVLQSI TDIIQINGIP TVVGVFTTQL NSIPGSAVCA FSMDDIEKVF KGRFKEQKTP DSVWTAVPED KVPKPRPGCC AKHGLAEAYK TSIDFPDETL SFIKSHPLMD SAVPPIADEP WFTKTRVRYR LTAISVDHSA GPYQNYTVIF VGSEAGMVLK VLAKTSPFSL NDSVLLEEIE AYNHAK
Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphoring domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. SEMA6D is a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11598; KIAA1479FLJ11598; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D; Semaphorin-6D
Gene Aliases: KIAA1479; SEMA6D
UniProt ID: (Human) Q8NFY4
Entrez Gene ID: (Human) 80031
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support