Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDDIADRMR MDAGEVTLVN HNSVFKTHLL PQTGFPEDQL SLSDQQILSS RQGHLDRSFT CSTRSAAYNP SYYSDNPSSD SFLGSGDLRT FGQSANGQWR NSTPSSSSSL QKSRNSRSLY LETRKTSSGL SNSFAGKSNH HCHVSAYEKS FPIKPVPSPS WSGSCRRSLL SPKKTQRRHV STAEETVQEE EREVYRQLLQ MVTGKQFTIA KPTTHFPLHL SRCLSSSKNT LKDSLFKNGN SCASQIIGSD TSSSGSASIL TNQEQLSHSV YSLSSYTPDV AFGSKDSGTL HHPHHHHSVP HQPDNLAASN TQSEGSDSVI LLKVKDSQTP TP (1-331 aa encoded by BC045639)
SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SEN1; Sentrin-specific protease 1; Sentrin/SUMO-specific protease SENP1; SUMO1/sentrin specific protease 1
Gene Aliases: SENP1; SuPr-2
UniProt ID: (Human) Q9P0U3
Entrez Gene ID: (Human) 29843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support