Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASSEVARHL LFQSHMATKT TCMSSQGSDD EQIKRENIRS LTMSGHVGFE SLPDQLVNRS IQQGFCFNIL CVGETGIGKS TLIDTLFNTN FEDYESSHFC PNVKLKAQTY ELQESNVQLK LTIVNTVGFG DQINKEESYQ PIVDYIDAQF EAYLQEELKI KRSLFTYHDS RIHVCLYFIS PTGHSLKTLD LLTMKNLDSK VNIIPVIAKA DTVSKTELQK FKIKLMSELV SNGVQIYQFP TDDDTIAKVN AAMNGQLPFA VVGSMDEVKV GNKMVKARQY PWGVVQVENE NHCDFVKLRE MLICTNMEDL REQTHTRHYE LYRRCKLEEM GFTDVGPENK PVSVQETYEA KRHEFHGERQ RKEEEMKQMF VQRVKEKEAI LKEAERELQA KFEHLKRLHQ EERMKLEEKR RLLEEEIIAF SKKKATSEIF HSQSFLATGS NLRKDKDRKN SNFL
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11619; Septin-10
Gene Aliases: SEPT10; SEPTIN10
UniProt ID: (Human) Q9P0V9
Entrez Gene ID: (Human) 151011
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support