Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YDLQESNVRL KLTIVSTVGF GDQINKEDSY KPIVEFIDAQ FEAYLQEELK IRRVLHTYHD SRIHVCLYFI APTGHSLKSL DLVTMKKLDS KVNIIPIIAK ADAISKSELT KFKIKITSEL VSNGVQIYQF PTDDESVAEI NGTMNAHLPF AVIGSTEELK IGNKMMRARQ YPWGTVQVEN EAHCDFVKLR EMLIRVNMED LREQTHTRHY ELYRRCKLEE MGFKDTDPDS KPFSLQETYE AKRNEFLGEL QKKEEEMRQM FVQRVKEKEA ELKEAEKELH EKFDRLKKLH QDEKKKLEDK KKSLDDEVNA FKQRKTAAEL LQSQGSQAGG SQTLKRDKEK KNNPWLCTE
SEPT6 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. One version of pediatric acute myeloid leukemia is the result of a reciprocal translocation between chromosomes 11 and X, with the breakpoint associated with the genes encoding the mixed-lineage leukemia and septin 2 proteins. This gene encodes four transcript variants encoding three distinct isoforms. An additional transcript variant has been identified, but its biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Sep; 6-Sep; MGC16619; MGC20339; RP5-876A24.2; septin 2; septin 6; Septin-6
Gene Aliases: 2810035H17Rik; C920001C06Rik; KIAA0128; mKIAA0128; SEP2; Sep6; SEPT2; SEPT6; SEPTIN6
UniProt ID: (Human) Q14141, (Mouse) Q9R1T4
Entrez Gene ID: (Rabbit) 100341047, (Human) 23157, (Mouse) 56526, (Rat) 691335
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support