Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AHKMKEHIPF FEDMCKGIKA GDTCEKLV
Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 79%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine incorporator 5
Gene Aliases: C5orf12; SERINC5; TPO1
UniProt ID: (Human) Q86VE9
Entrez Gene ID: (Human) 256987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support