Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Mouse Serpinb3c.
SPB3 may act as a protease inhibitor to modulate the host immune response against tumor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: clade B (ovalbumin); ovalbumin; Protein T4-A; SCCA; SCCA-1; SCCA1; serine (or cysteine) proteinase inhibitor; serine (or cysteine) proteinase inhibitor, clade B, member 3C; Serpin B3; serpin peptidase inhibitor; SERPINB3; spb3; Squamous cell carcinoma antigen 1
Gene Aliases: 1110001H02Rik; 1110013A16Rik; Scca2; Serpinb4
Entrez Gene ID: (Mouse) 381286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support