Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
SERPINB4 is also called squamous cell carcinoma antigen 2 (SCCA-2) or leupin. It directly inhibits human granzyme proteolytic activity and that overexpression of SERPINB4 in HeLa cells inhibits recombinant granzyme-induced as well as NK cell-mediated cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leupin; Leupin SERPINB4; Peptidase inhibitor 11; PI-11; protease inhibitor (leucine-serpin); SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; Serpin B4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; squamous cell carcinoma antigen 1; Squamous cell carcinoma antigen 2
Gene Aliases: LEUPIN; PI11; SCCA-2; SCCA1; SCCA2; SERPINB4
UniProt ID: (Human) P48594
Entrez Gene ID: (Human) 6318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support