Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SERPINE2 Antibodies

          Abnova

          SERPINE2 Polyclonal Antibody, MaxPab™

          View all (39) SERPINE2 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SERPINE2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          SERPINE2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SERPINE2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SERPINE2 Antibody (H00005270-B01P) in WB

          Western Blot analysis of SERPINE2 expression in transfected 293T cell line by SERPINE2 MaxPab polyclonal antibody. Lane 1: SERPINE2 transfected lysate(44.00 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SERPINE2 Antibody in Western Blot (WB)

          Product Details

          H00005270-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SERPINE2 (NP_006207.1, 1 a.a. approximately 398 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MNWHLPLFLL ASVTLPSICS HFNPLSLEEL GSNTGIQVFN QIVKSRPHDN IVISPHGIAS VLGMLQLGAD GRTKKQLAMV MRYGVNGVGK ILKKINKAIV SKKNKDIVTV ANAVFVKNAS EIEVPFVTRN KDVFQCEVRN VNFEDPASAC DSINAWVKNE TRDMIDNLLS PDLIDGVLTR LVLVNAVYFK GLWKSRFQPE NTKKRTFVAA DGKSYQVPML AQLSVFRCGS TSAPNDLWYN FIELPYHGES ISMLIALPTE SSTPLSAIIP HISTKTIDSW MSIMVPKRVQ VILPKFTAVA QTDLKEPLKV LGITDMFDSS KANFAKITTG SENLHVSHIL QKAKIEVSED GTKASAATTA ILIARSSPPW FIVDRPFLFF IRHNPTGAVL FMGQINKP

          Target Information

          This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GDN; Glia derived nexin; Glia-derived nexin; glial-derived neurite promoting factor; P17; Peptidase inhibitor 7; PI-7; PN-1; Protease nexin 1; Protease nexin I; Serpin E2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; SERPINE 2

          View more View less

          Gene Aliases: GDN; GDNPF; PI-7; PI7; PN-1; PN1; PNI; SERPINE2

          View more View less

          UniProt ID: (Human) P07093

          View more View less

          Entrez Gene ID: (Human) 5270

          View more View less

          Function(s)
          serine-type endopeptidase inhibitor activity receptor binding protein binding glycosaminoglycan binding heparin binding
          Process(es)
          embryo implantation blood coagulation negative regulation of cell proliferation response to radiation negative regulation of plasminogen activation negative regulation of sodium ion transport negative regulation of endopeptidase activity negative regulation of protein processing positive regulation of neuron projection development negative regulation of phosphatidylinositol 3-kinase signaling cerebellar granular layer morphogenesis negative regulation of blood coagulation negative regulation of cell growth regulation of cell migration secretion by cell secretory granule organization negative regulation of protein catabolic process mating plug formation negative regulation of proteolysis negative regulation of smoothened signaling pathway regulation of timing of cell differentiation positive regulation of astrocyte differentiation detection of mechanical stimulus involved in sensory perception regulation of synaptic transmission, glutamatergic long-term synaptic potentiation innervation seminal vesicle epithelium development dense core granule biogenesis negative regulation of platelet aggregation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0