Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MQALVLLLCI GALLGHSSCQ NPASPPEEGS PDPDSTGALV EEEDPFFKVP VNKLAAAVSN FGYDLYRVRS STSPTTNVLL SPLSVATALS ALSLGAEQRT ESIIHRALYY DLISSPDIHG TYKELLDTVT APQKNLKSAS RIVFEKKLRI KSSFVAPLEK SYGTRPRVLT GNPRLDLQEI NNWVQAQMKG KLARSTKEIP DEISILLLGV AHFKGQWVTK FDSRKTSLED FYLDEERTVR VPMMSDPKAV LRYGLDSDLS CKIAQLPLTG SMSIIFFLPL KVTQNLTLIE ESLTSEFIHD IDRELKTVQA VLTVPKLKLS YEGEVTKSLQ EMKLQSLFDS PDFSKITGKP IKLTQVEHRA GFEWNEDGAG TTPSPGLQPA HLTFPLDYHL NQPFIFVLRD TDTGALLFIG KILDPRGP
PEDF is a noninhibitory serpin with neurotrophic, anti-angiogenic, and anti-tumorigenic properties. It is a 50 kDa glycoprotein produced and secreted in many tissues throughout the body. A major component of the anti-angiogenic action of PEDF is the induction of apoptosis in proliferating endothelial cells. In addition, PEDF is able to inhibit the activity of angiogenic factors, such as VEGF and FGF-2. The neuroprotective effects of PEDF are achieved through suppression of neuronal apoptosis induced by peroxide, glutamate, or other neurotoxins. The recent identification of a lipase-linked cell membrane receptor for PEDF (PEDF-R) that binds to PEDF with high affinity (1) should facilitate further elucidation of the underlying mechanisms of this pluripotent serpin. To date, PEDF-R is the only signaling receptor known to be used by a serpin family member. The unique range of PEDF activities implicate it as a potential therapeutic agent for the treatment of vasculature-related neurodegenerative diseases, such as age-related macular degeneration (AMD) and proliferative diabetic retinopathy (PDR). PEDF also has the potential to be useful in the treatment of various angiogenesis-related diseases including a number of cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 35 protein; EPC 1; EPC-1; EPC1; PEDF; PIG 35; Pigment epithelium-derived factor; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; Serpin F1; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; SERPINF 1; sserpin family F member 1; testis tissue sperm-binding protein Li 70n
Gene Aliases: EPC-1; OI12; OI6; PEDF; PIG35; SERPINF1
UniProt ID: (Human) P36955
Entrez Gene ID: (Human) 5176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support