Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLSKGLKRKR EEEEEKEPLA VDSWWLDPGH TAVAQAPPAV ASSSLFDLSV LKLHHSPQQS EPDLRHLVLV VNTLRRIQAS MAPAAALPPV PSPPAAPSVA DNLLASSDAA LSASMASLLE DLSHIEGLSQ APQPLADEGP PGRSIGGAAP SLGALDLLGP ATGCLLDDGL EGLFEDIDTS MYDNELWAPA SEGLKPGPED GPGKEEAPEL DEAELDYLMD VLVGTQALER PPGPGR
SEI-1, also known as SERTAD1 (SERTA domain containing 1) or TRIP-Br1 (Transcriptional regulator interacting with the PHD-bromodomain 1), is a transcriptional regulator that integrates signals provided by transcription factors. Acting at E2F-responsive promoters, SEI-1 interacts with the PHD- and bromodomains of proteins such as TIF1 and DP-1, thereby transmitting their signals to the promoter and stimulating transcriptional activity. SEI-1 exists as a multiprotein complex with E2F-1 and DP-1 and is expressed at different levels throughout the cell cycle, allowing it to regulate cell cycle progression via promoter control during the G1 and S phases. Additionally, SEI-1 can render the activity of the cyclin D-Cdk4 complex, an important catalyst of the cell cycle, resistant to the inhibitory effects of p16. Overexpression of SEI-1 is implicated in the development of squamous cell carcinomas of the head and neck.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK4-binding protein p34SEI; CDK4-binding protein p34SEI1; SEI-1; SERTA domain-containing protein 1; Transcriptional regulator interacting with the PHD-bromodomain 1; TRIP-Br1
Gene Aliases: SEI1; SERTAD1; TRIP-Br1; TRIPBR1
UniProt ID: (Human) Q9UHV2
Entrez Gene ID: (Human) 29950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support