Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDDPSVGPKI PQALEKILQL KESRQEEMNS QQEEEEMETD ARSSLGQSAS ETEEDTVSVS KKEKNRKRRN RKKKKKPQRV RGVSSESSGD REKDSTRSRG SDSPAADVEI EYVTEEPEIY EPNFIFFKRI FEAFKLTDDV KKEKEKEPEK LDKLENSAAP KKKGFEEEHK DSDDDSSDDE QEKKPEAPKL SKKKLRRMNR FTVAELKQLV ARPDVVEMHD VTAQDPKLLV HLKATRNSVP VPRHWCFKRK YLQGKRGIEK PPFELPDFIK RTGIQEMREA LQEKEEQKTM KSKMREKVRP KMGKIDIDYQ KLHDAFFKWQ TKPKLTIHGD LYYEGKEFET RLKEKKPGDL SDELRISLGM PVGPNAHKVP PPWLIAMQRY GPPPSYPNLK IPGLNSPIPE SCSFGYHAGG WGKPPVDETG KPLYGDVFGT NAAEFQTKTE EEEIDRTPWG ELEPSDEESS EEEEEEESDE DKPDETGFIT PADSGLITPG GFSSVPAGME TPELIELRKK KIEEAMDGSE TPQLFTVLPE KRTATVGGAM MGSTHIYDMS TVMSRKGPAP ELQGVEVALA PEELELDPMA MTQKYEEHVR EQQAQVEKED FSDMVAEHAA KQKQKKRKAQ PQDSRGGSKK YKEFKF
Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pre-mRNA splicing factor SF3b 145 kDa subunit; Pre-mRNA-splicing factor SF3b 145 kDa subunit; SAP 145; SF3b145; spliceosome associated protein 145; Spliceosome-associated protein 145; Splicing factor 3B subunit 2; splicing factor 3b, subunit 2, 145kD; splicing factor 3b, subunit 2, 145kDa
Gene Aliases: Cus1; SAP145; SF3b1; SF3B145; SF3b150; SF3B2
UniProt ID: (Human) Q13435
Entrez Gene ID: (Human) 10992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support