Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SFRS15 Antibodies

          Abnova

          SFRS15 Polyclonal Antibody, MaxPab™

          View all (9) SFRS15 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SFRS15 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          SFRS15 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SFRS15 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SFRS15 Antibody (H00057466-B01P) in WB

          Western Blot analysis of SFRS15 expression in transfected 293T cell line by SFRS15 MaxPab polyclonal antibody. Lane 1: SFRS15 transfected lysate(126.17 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SFRS15 Antibody in Western Blot (WB)

          Product Details

          H00057466-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SFRS15 (NP_065757.1, 1 a.a. approximately 1147 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDAVNAFNQE LFSLMDMKPP ISRAKMILIT KAAIKAIKLY KHVVQIVEKF IKKCKPEYKV PGLYVIDSIV RQSRHQFGTD KDVFGPRFSK NITATFQYLY LCPSEDKSKI VRVLNLWQKN GVFKIEIIQP LLDMAAGTSN AAPVAENVTN NEGSPPPPVK VSSEPPTQAT PNSVPAVPQL PSSDAFAAVA QLFQTTQGQQ LQQILQTFQQ PPKPQSPALD NAVMAQVQAI TAQLKTTPTQ PSEQKAAFPP PEQKTAFDKK LLDRFDYDDE PEAVEESKKE DTTAVTTTAP AAAVPPAPTA TVPAAAAPAA ASPPPPQAPF GFPGDGMQQP AYTQHQNMDQ FQPRMMGIQQ DPMHHQVPLP PNGQMPGFGL LPTPPFPPMA QPVIPPTPPV QQPFQASFQA QNEPLTQKPH QQEMEVEQPC IQEVKRHMSD NRKSRSRSAS RSPKRRRSRS GSRSRRSRHR RSRSRSRDRR RHSPRSRSQE RRDREKERER RQKGLPQVKP ETASVCSTTL WVGQLDKRTT QQDVASLLEE FGPIESINMI PPRGCAYIVM VHRQDAYRAL QKLSRGNYKV NQKSIKIAWA LNKGIKADYK QYWDVELGVT YIPWDKVKPE ELESFCEGGM LDSDTLNPDW KGIPKKPENE VAQNGGAETS HTEPVSPIPK PLPVPVPPIP VPAPITVPPP QVPPHQPGPP VVGALQPPAF TPPLGIPPPG FGPGVPPPPP PPPFLRPGFN PMHLPPGFLP PGPPPPITPP VSIPPPHTPP ISIPNSTIAG INEDTTKDLS IGNPIPTVVS GARGNAESGD SVKMYGSAVP PAAPTNLPTP PVTQPVSLLG TQGVAPGPVI GLQAPSTGLL GARPGLIPLQ RPPGMPPPHL QRFPLMPPRP MPPHMMHRGP PPGPGGFAMP PPHGMKGPFP PHGPFVRPGG MPGLGGPGPG PGGPEDRDGR QQPPQQPQQQ PQPQAPQQPQ QQQQQQPPPS QQPPPTQQQP QQFRNDNRQQ FNSGRDQERF GRRSFGNRVE NDRERYGNRN DDRDNSNRDR REWGRRSPDR DRHRDLEERN RRSSGHRDRE RDSRDRESRR EKEEARGKEK PEVTDRAGGN KTVEPPISQV GNVDTASELE KGVSEAAVLK PSEELPAEAT SSVEPEKDSG SAAEAPR

          Target Information

          The family of SR factors all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. They are not only essential for constitutive splicing, but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Splicing factor arginine/serine-rich 15 (SFRS15), also designated CTD-binding SR-like protein RA4, contains one RRM and one SR-rich domains. SFRS15 interacts with C-terminal repetitive domain (CTD) of Pol II and is believed to functionally and physically link transcription and pre-mRNA processing. Localized to the nucleus, SFRS15 is expressed as two isoforms produced by alternative splicing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CTD-binding SR-like protein RA4; pre-mRNA splicing SR protein rA4; SFR15; splicing factor serine alanine 15; Splicing factor, arginine/serine-rich 15; SR-like CTD-associated factor 4; SR-related and CTD-associated factor 4

          View more View less

          Gene Aliases: KIAA1172; SCAF4; SFRS15; SRA4

          View more View less

          UniProt ID: (Human) O95104

          View more View less

          Entrez Gene ID: (Human) 57466

          View more View less

          Function(s)
          nucleotide binding poly(A) RNA binding
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0