Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17D9; Caspase-11; Caspase11; CTD-associated SR protein 11; p30 subunit; Protein SCAF11; Renal carcinoma antigen NY-REN-40; SC35-interacting protein 1; serine/arginine-rich splicing factor 2, interacting protein; Serine/arginine-rich splicing factor 2-interacting protein; SFRS2-interacting protein; splicing factor, arginine/serine-rich 2, interacting protein; Splicing factor, arginine/serine-rich 2-interacting protein; Splicing regulatory protein 129; SR-related and CTD-associated factor 11; SR-related CTD-associated factor 11; SRrp129; SRSF2-interacting protein
Gene Aliases: CASP11; SCAF11; SFRS2IP; SIP1; SRRP129; SRSF2IP
UniProt ID: (Human) Q99590
Entrez Gene ID: (Human) 9169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support