Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha h UBP; Alpha-SGT; protein containing three tetratricopeptide repeats; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; Small glutamine-rich tetratricopeptide repeat-containing protein alpha; UBP; Vpu-binding protein
Gene Aliases: 5330427H01Rik; AI194281; alphaSGT; D10Ertd190e; hSGT; SGT; SGT1; SGTA; Stg
UniProt ID: (Human) O43765, (Mouse) Q8BJU0
Entrez Gene ID: (Human) 6449, (Mouse) 52551
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support