Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEFPLAQICP QGSHEAPIPT FSTFQITDMT RRSCQNLGYT AASPQAPEAA SSTGNAERAE EVPGEGSLFL QAETRAWFQK TQAHWLLQHG AAPAWFHGFI TRREAERLLE PKPQGCYLVR FSESAVTFVL TYRSRTCCRH FLLAQLRDGR HVVLGEDSAH ARLQDLLLHY TAHPLSPYGE TLTEPLARQT PEPAGLSLRT EESNFGSKSQ DPNPQYSPII KQGQAPVPMQ KEGAGEKEPS QLLRPKPPIP AKPQLPPEVY TIPVPRHRPA PRPKPSNPIY NEPDEPIAFY AMGRGSPGEA PSNIYVEVED EGLPATLGHP VLRKSWSRPV PGGQNTGGSQ LHSENSVIGQ GPPLPHQPPP AWRHTLPHNL SRQVLQDRGQ AWLPLGPPQ
Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH2 domain protein 2A; SH2 domain-containing adapter protein; SH2 domain-containing protein 2A; SH22A; T cell specific adapter protein TSAd; T cell-specific adapter protein; T lymphocyte specific adaptor protein; TSAd; VEGF receptor-associated protein
Gene Aliases: F2771; SCAP; SH2D2A; TSAD; VRAP
UniProt ID: (Human) Q9NP31
Entrez Gene ID: (Human) 9047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support