Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTAVGRRCPA LGSRGAAGEP EAGSDYVKFS KEKYILDSSP EKLHKELEEE LKLSSTDLRS HAWYHGRIPR EVSETLVQRN GDFLIRDSLT SLGDYVLTCR WRNQALHFKI NKVVVKAGES YTHIQYLFEQ ESFDHVPALV RYHVGSRKAV SEQSGAIIYC PVNRTFPLRY LEASYGLGQG SSKPASPVSP SGPKGSHMKR RSVTMTDGLT ADKVTRSDGC PTSTSLPRPR DSIRSCALSM DQIPDLHSPM SPISESPSSP AYSTVTRVHA APAAPSATAL PASPVARRSS EPQLCPGSAP KTHGESDKGP HTSPSHTLGK ASPSPSLSSY SDPDSGHYCQ LQPPVRGSRE WAATETSSQQ ARSYGERLKE LSENGAPEGD WGKTFTVPIV EVTSSFNPAT FQSLLIPRDN RPLEVGLLRK VKELLAEVDA RTLARHVTKV DCLVARILGV TKEMQTLMGV RWGMELLTLP HGRQLRLDLL ERFHTMSIML AVDILGCTGS AEERAALLHK TIQLAAELRG TMGNMFSFAA VMGALDMAQI SRLEQTWVTL RQRHTEGAIL YEKKLKPFLK SLNEGKEGPP LSNTTFPHVL PLITLLECDS APPEGPEPWG STEHGVEVVL AHLEAARTVA HHGGLYHTNA EVKLQGFQAR PELLEVFSTE FQMRLLWGSQ GASSSQARRY EKFDKVLTAL SHKLEPAVRS SEL
Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas/HEF1-associated signal transducer; Chat-H; Novel SH2-containing protein 3; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; SHEP1
Gene Aliases: CHAT; NSP3; PRO34088; SH2D3C; SHEP1; UNQ272/PRO309/PRO34088
UniProt ID: (Human) Q8N5H7
Entrez Gene ID: (Human) 10044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support